Zinc protoporphyrin guanylate cyclase inhibitor, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX

M-Clarity™ Qualitätsstufe = MQ100

282820-50MG 282820-250MG
282820-50MGEA 249 EUR
282820-50MG 282820-250MG
Zinc protoporphyrin guanylate cyclase inhibitor, Sigma-Aldrich®
Zinc protoporphyrin

Inhibits soluble guanylate cyclase.


Protoporphyrin IX zinc(II) (ZnPP) is an anti-oxidant enzyme heme oxygenase-1. Cellular melanin content in normal human melanocytes treated with ZnPP has been studied. 1 It was used as a standard in a study where the excess formation of ZnPP in iron deficient erythropoiesis was determined by haematofluorometer.

Formel: C₃₄H₃₂N₄O₄Zn
Molecular Weight: 626,034 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00011612
CAS-Nummer: 15442-64-5
EINECS: 239-455-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29508150f149e320fdcafa43ce2076ffbd901772&vid=30fd600ac8c511994c7176d4211360c35aa2e9aa&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Red to Very Dark Red and Purple to Very Dark Purple
Appearance (Form) Powder or Crystals
Infrared spectrum, C=0.005 G/L Pyridine 585 - 591 nm Conforms to Structure
Extinction Coefficient ≥10000
Wavelength 423 - 429 nm
Extinction Coefficient, In Pyridine ≥135000
Carbon 58.7 - 71.8%
Nitrogen 8.1 - 9.8%
Purity (HPLC) ≥92.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR