5-Aminofluorescein (isomer I), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one (isomer I), 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-Aminofluorescein, Fluoresceinamine, isomer I, 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine (isomer I)

M-Clarity™ Qualitätsstufe = MQ200

201626-1G 201626-250MG 201626-5G
201626-1GEA 186 EUR
201626-1G 201626-250MG 201626-5G
5-Aminofluorescein (isomer I), Sigma-Aldrich®
5-Aminofluorescein (isomer I)

Fluoresceinamine, isomer I has been used as a probe in immobilization and is used to determine its ability to potentiate the antiviral activity of poly r (A-U).


Fluoresceinamine, isomer I belongs to the class of derivatized fluoresceins. Fluoresceinamine, isomer I is suitable for use in a specific and sensitive spectrophotometric method for determining nitrite. It has been used to fluorescently tag nanoparticles through a competitive carboxyl-amine coupling reaction to visualise nanoparticle internalisation.

Formel: C₂₀H₁₃NO₅
Molecular Weight: 347,33 g/mol
Schmelzpunkt: 223 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00005052
CAS-Nummer: 3326-34-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7a811f20096b5c5331ed556e43f05c852a310e5&vid=a7a811f20096b5c5331ed556e43f05c852a310e5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon, Anhydrous Basis 67.8 - 70.6%
Nitrogen, Anhydrous Basis 3.7 - 4.3%
Purity (HPLC) Typically 95+%
Water (by Karl Fischer) ≤10%
Solubility (Colour), c = 5 mg/ml (0.5%); Methanol Yellow to Very Dark Yellow and Orange to Very Dark Orange
Recommended Retest Period 4 Years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR