1-(2-Hydroxyethyl)-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 1,2,3,4-Tetrahydro-2,2,4,7-tetramethyl-1-quinolineethanol

578355-100G 578355-25G
578355-100GEA 381 EUR
578355-100G 578355-25G
1-(2-Hydroxyethyl)-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline 97%, Sigma-Aldrich®
1-(2-Hydroxyethyl)-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline

1-(2-Hydroxyethyl)-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline,97% (1,2,3,4-Tetrahydro-2,2,4,7-tetramethyl-1-quinolineethanol)

Formel: C₁₅H₂₃NO
Molecular Weight: 233.35 g/mol
Schmelzpunkt: 96…100 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00155229
CAS-Nummer: 53817-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8a2f39576f85c35f225ac3b513752b79c827618&vid=56288b3660bbd2dabe2b9dca6b66940447e296bc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR