4-Pentylbicyclo[2.2.2]octane-1-carboxylic acid 99%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

393363-1G 393363-5G
393363-1GEA 74 EUR
393363-1G 393363-5G
4-Pentylbicyclo[2.2.2]octane-1-carboxylic acid 99%, Sigma-Aldrich®
4-Pentylbicyclo[2.2.2]octane-1-carboxylic acid

4-Pentylbicyclo[2.2.2]octane-1-carboxylic acid, 99%.

Formel: C₁₄H₂₄O₂
Molecular Weight: 224,34 g/mol
Schmelzpunkt: 160,..162 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00192383
CAS-Nummer: 73152-70-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f31d100b1ba9b66bff17d1cd37290f940c9df82&vid=5f31d100b1ba9b66bff17d1cd37290f940c9df82&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Off-White
Appearance (Form): Powder or Crystals Conforms to Requirements
Infrared spectrum Conforms to Structure
Purity (Titration by NaOH) 98.5 - 101.5%
GC (area %) ≥98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR