α-Ketoglutaric acid disodium salt dihydrate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Oxoglutaric acid disodium salt dihydrate, α-Ketoglutaric acid disodium salt dihydrate, Sodium α-ketoglutarate dibasic dihydrate
BI2993-100G
BI2993-25G
BI2993-50G
APOSBI2993-100GEA
125
EUR
APOSBI2993-100G
APOSBI2993-25G
APOSBI2993-50G
α-Ketoglutaric acid disodium salt dihydrate
α-Ketoglutaric acid disodium salt dihydrate
Assay 82+% Forms a clear solution in water (1.0M)
- Biological buffer
Formel:
C₅H₄O₅·2Na·2H₂O Molecular Weight: 226,092 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00150702 CAS-Nummer: 1282616-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...