5-Bromo-4-chloro-3-indolyl phenyl phosphonate, p-toluidine salt
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
X-Pphos, XPP
BI5949-100MG
BI5949-250MG
APOSBI5949-100MGEA
215
EUR
APOSBI5949-100MG
APOSBI5949-250MG
5-Bromo-4-chloro-3-indolyl phenyl phosphonate, p-toluidine salt
5-Bromo-4-chloro-3-indolyl phenyl phosphonate, p-toluidine salt
Chromogenic enzyme substrate for the detection ofphosphonate monoesterase, a conditional lethal gene (pehA) used in the study of plant genetics.