L-Aminoxy-3-phenylpropionic acid hydrobromide
Lieferant: Apollo Scientific
Synonyme: (2S)-2-(Aminooxy)-3-phenylpropanoic acid hydrobromide
BIA1399-100MG BIA1399-25MG
APOSBIA1399-100MGEA 625 EUR
APOSBIA1399-100MG APOSBIA1399-25MG
L-Aminoxy-3-phenylpropionic acid hydrobromide
L-Aminoxy-3-phenylpropionic acid hydrobromide
Formel: C₉H₁₁NO₃·BrH
Molecular Weight: 262,1 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00269699
CAS-Nummer: 73086-97-2