15-Acetyl-deoxynivalenol 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 15ADON, 15-Acetyl Deoxynivalenol, 15-O-Acetyl-4-deoxynivalenol, 15-acetyl don, 15-Acetoxy-3α,7α-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 15-Acetoxy-3-α,7-α-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 15-acetyldeoxynivalenol

BIA2302-1MG BIA2302-5MG
APOSBIA2302-1MGEA 364 EUR
APOSBIA2302-1MG APOSBIA2302-5MG
15-Acetyl-deoxynivalenol 99%
15-Acetyl-deoxynivalenol

From Fusarium, mycotoxin, protein synthesis inhibitor

Formel: C₁₇H₂₂O₇
Molecular Weight: 338,352 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00083218
CAS-Nummer: 88337-96-6
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR