6-Bromo-2-naphthyl-β-D-galactopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-2-naphthyl b-D-galactoside, 6-Bromo-2-naphthyl β-D-galactopyranoside

BIB1201-1G BIB1201-500MG
APOSBIB1201-1GEA 116 EUR
APOSBIB1201-1G APOSBIB1201-500MG
6-Bromo-2-naphthyl-β-D-galactopyranoside
6-Bromo-2-naphthyl-β-D-galactopyranoside
Formel: C₁₆H₁₇BrO₆
Molecular Weight: 385,207 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00064182
CAS-Nummer: 15572-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR