5-Bromo-6-chloro-3-indolyl β-D-glucopyranoside
Lieferant: Apollo Scientific
Achtung
Synonyme: Magenta-glucoside, (2S,3R,4S,5S,6R)-2-((5-Bromo-6-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
BIB1417-1G BIB1417-250MG BIB1417-500MG
APOSBIB1417-1GEA 116 EUR
APOSBIB1417-1G APOSBIB1417-250MG APOSBIB1417-500MG
5-Bromo-6-chloro-3-indolyl β-D-glucopyranoside
5-Bromo-6-chloro-3-indolyl β-D-glucopyranoside
Formel: C₁₄H₁₅BrClNO₆
Molecular Weight: 408.629 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00210022
CAS-Nummer: 93863-89-9