4-Nitrophenyl α-D-xylopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,3R,4S,5R)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

BIB1435-100MG BIB1435-250MG BIB1435-50MG
APOSBIB1435-100MGEA 102 EUR
APOSBIB1435-100MG APOSBIB1435-250MG APOSBIB1435-50MG
4-Nitrophenyl α-D-xylopyranoside
4-Nitrophenyl α-D-xylopyranoside
Formel: C₁₁H₁₃NO₇
Molecular Weight: 271.223 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00067649
CAS-Nummer: 10238-28-5
EINECS: 233-572-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR