5-Chloro-6-methoxyindole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-6-methoxy-1H-indole

BIB6030-1G BIB6030-250MG
APOSBIB6030-1GEA 154 EUR
APOSBIB6030-1G APOSBIB6030-250MG
5-Chloro-6-methoxyindole
5-Chloro-6-methoxyindole
Formel: C₉H₈ClNO
Molecular Weight: 181,619 g/mol
Schmelzpunkt: 109…110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04972054
CAS-Nummer: 90721-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR