D-Glucuronic acid, sodium salt monohydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: Sodium D-glucuronate

BIB6055-100G BIB6055-250G BIB6055-500G
APOSBIB6055-100GEA 122 EUR
APOSBIB6055-100G APOSBIB6055-250G APOSBIB6055-500G
D-Glucuronic acid, sodium salt monohydrate
D-Glucuronic acid, sodium salt monohydrate
Formel: C₆H₉O₇·Na·H₂O
Molecular Weight: 234.137 g/mol
Schmelzpunkt: 138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151051
CAS-Nummer: 14984-34-0
EINECS: 239-065-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR