5-Hydroxytryptophol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(2-Hydroxyethyl)-1H-indol-5-ol

BIB6072-1G BIB6072-250MG
APOSBIB6072-1GEA 290 EUR
APOSBIB6072-1G APOSBIB6072-250MG
5-Hydroxytryptophol
5-Hydroxytryptophol
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,1998 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00047197
CAS-Nummer: 154-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR