5-Iodo-3-indolyl-β-D-galactopyranoside
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Purple β-D-gal
BIB6084-100MG
BIB6084-500MG
BIB6084-250MG
APOSBIB6084-100MGEA
116
EUR
APOSBIB6084-100MG
APOSBIB6084-500MG
APOSBIB6084-250MG
5-Iodo-3-indolyl-β-D-galactopyranoside
5-Iodo-3-indolyl-β-D-galactopyranoside
Formel:
C₁₄H₁₆INO₆ Molecular Weight: 421,184 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00798400 CAS-Nummer: 36473-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...