2-Naphthyl caprylate
Lieferant: Apollo Scientific
|
Achtung
|
BIB6122-1G
BIB6122-500MG
APOSBIB6122-1GEA
270
EUR
APOSBIB6122-1G
APOSBIB6122-500MG
2-Naphthyl caprylate
2-Naphthyl caprylate
Lagertemperatur:
Kühlschrank |
MDL:
MFCD00046465 CAS-Nummer: 10251-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...