4-Nitrophenyl-α-D-maltohexaoside

Lieferant: Apollo Scientific
Achtung

BIB6130-1G BIB6130-250MG BIB6130-500MG
APOSBIB6130-1GEA 719 EUR
APOSBIB6130-1G APOSBIB6130-250MG APOSBIB6130-500MG
4-Nitrophenyl-α-D-maltohexaoside
4-Nitrophenyl-α-D-maltohexaoside
Lagertemperatur: Kühlschrank
MDL: MFCD00065704
CAS-Nummer: 74173-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR