4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt

Lieferant: Apollo Scientific

Synonyme: 4-Nitrophenyl phosphate bis(cyclohexylammonium) salt

BIB6134-10G BIB6134-50G BIB6134-5G
APOSBIB6134-10GEA 284 EUR
APOSBIB6134-10G APOSBIB6134-50G APOSBIB6134-5G
4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt
4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt
Formel: C₆H₆NO₆P·2C₆H₁₃N
Molecular Weight: 417,437 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00013173
CAS-Nummer: 52483-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR