N-Tosyl-L-alanine 3-indoxyl ester
Lieferant: Apollo Scientific
|
Achtung
|
BIB6276-1G
BIB6276-5G
APOSBIB6276-1GEA
241
EUR
APOSBIB6276-1G
APOSBIB6276-5G
N-Tosyl-L-alanine 3-indoxyl ester
N-Tosyl-L-alanine 3-indoxyl ester
Formel:
C₁₈H₁₈N₂O₄S Molecular Weight: 358.412 g/mol Schmelzpunkt: 77 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03701236 CAS-Nummer: 75062-54-3 EINECS: 278-064-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...