Acedoben-acyl-β-D-glucuronide ≥98%
Lieferant: Apollo Scientific
Synonyme: PADB-β-D-glucuronide, 1-[4-(Acetylamino)benzoate]-β-D-glucopyranuronic acid, 1-(p-Acetamidobenzoate)-β-D-glucopyranuronic acid
BICL4005-10MG BICL4005-25MG BICL4005-5MG
APOSBICL4005-10MGEA 400 EUR
APOSBICL4005-10MG APOSBICL4005-25MG APOSBICL4005-5MG
Acedoben-acyl-β-D-glucuronide ≥98%
Acedoben-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 34220-56-9