2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3,4,6-Tetra-O-acetyl-GlcNAc, D-Glucosamine pentaacetate, 1,3,4,6-Tetra-O-acetyl-2-(acetylamino)-2-deoxy-D-glucopyranose, (3R,4R,5S,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate, GlcNAc tetraacetate
BICL4009-10G
BICL4009-25G
BICL4009-50G
APOSBICL4009-10GEA
422
EUR
APOSBICL4009-10G
APOSBICL4009-25G
APOSBICL4009-50G
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose ≥99%
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose
Formel:
C₁₆H₂₃NO₁₀ Molecular Weight: 389,3600 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00067367 CAS-Nummer: 14086-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...