1,6-Anhydro-3-O-benzyl-β-L-idopyranose ≥98%
Lieferant: Apollo Scientific
BICL4028-100MG
BICL4028-500MG
APOSBICL4028-100MGEA
290
EUR
APOSBICL4028-100MG
APOSBICL4028-500MG
1,6-Anhydro-3-O-benzyl-β-L-idopyranose ≥98%
1,6-Anhydro-3-O-benzyl-β-L-idopyranose
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
42926-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...