Benzyl 2-acetamido-2-deoxy-4,6-O-isopropylidene-β-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzyl 2-(acetylamino)-2-deoxy-4,6-O-isopropylidene-β-D-glucopyranoside, Benzyl 4,6-O-isopropylidene-β-GlcNAc, BADIG

BICL4041-1G BICL4041-2G
APOSBICL4041-1GEA 517 EUR
APOSBICL4041-1G APOSBICL4041-2G
Benzyl 2-acetamido-2-deoxy-4,6-O-isopropylidene-β-D-glucopyranoside ≥98%
Benzyl 2-acetamido-2-deoxy-4,6-O-isopropylidene-β-D-glucopyranoside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 50605-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR