Benzyl 2,3:4,6-di-O-isopropylidene-α-L-sorbofuranoside ≥98%

Lieferant: Apollo Scientific

BICL4052-1G BICL4052-500MG BICL4052-250MG
APOSBICL4052-1GEA 1200 EUR
APOSBICL4052-1G APOSBICL4052-500MG APOSBICL4052-250MG
Benzyl 2,3:4,6-di-O-isopropylidene-α-L-sorbofuranoside ≥98%
Benzyl 2,3:4,6-di-O-isopropylidene-α-L-sorbofuranoside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 74342-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR