Celecoxib carboxylic acid-acyl-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4055-1MG BICL4055-2MG
APOSBICL4055-1MGEA 517 EUR
APOSBICL4055-1MG APOSBICL4055-2MG
Celecoxib carboxylic acid-acyl-β-D-glucuronide ≥95%
Celecoxib carboxylic acid-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 264236-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR