D-Cellotriose undecaacetate ≥98%

Lieferant: Apollo Scientific

BICL4057-100MG BICL4057-25MG BICL4057-50MG
APOSBICL4057-100MGEA 1360 EUR
APOSBICL4057-100MG APOSBICL4057-25MG APOSBICL4057-50MG
D-Cellotriose undecaacetate ≥98%
D-Cellotriose undecaacetate
Lagertemperatur: Raumtemperatur
CAS-Nummer: 17690-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR