Diflunisal-acyl-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4063-1MG BICL4063-5MG BICL4063-2MG
APOSBICL4063-1MGEA 393 EUR
APOSBICL4063-1MG APOSBICL4063-5MG APOSBICL4063-2MG
Diflunisal-acyl-β-D-glucuronide ≥95%
Diflunisal-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 58446-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR