1,6:2,3-Dianhydro-4-O-benzyl-β-D-mannopyranose ≥99%

Lieferant: Apollo Scientific

BICL4082-100MG BICL4082-250MG BICL4082-50MG
APOSBICL4082-100MGEA 504 EUR
APOSBICL4082-100MG APOSBICL4082-250MG APOSBICL4082-50MG
1,6:2,3-Dianhydro-4-O-benzyl-β-D-mannopyranose ≥99%
1,6:2,3-Dianhydro-4-O-benzyl-β-D-mannopyranose
Lagertemperatur: Raumtemperatur
CAS-Nummer: 33208-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR