1,2-Di-O-acetyl-3,5-di-O-benzoyl-β-D-ribofuranose ≥99%
Lieferant: Apollo Scientific
BICL4088-100MG
BICL4088-250MG
APOSBICL4088-100MGEA
434
EUR
APOSBICL4088-100MG
APOSBICL4088-250MG
1,2-Di-O-acetyl-3,5-di-O-benzoyl-β-D-ribofuranose ≥99%
1,2-Di-O-acetyl-3,5-di-O-benzoyl-β-D-ribofuranose
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
71080-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...