2-(Dimethoxymethyl)-5-(methoxymethyl)furan ≥99%

Lieferant: Apollo Scientific

Synonyme: 5-(Methoxymethyl)-2-furaldehyde dimethyl acetal

BICL4091-100MG BICL4091-250MG BICL4091-50MG
APOSBICL4091-100MGEA 532 EUR
APOSBICL4091-100MG APOSBICL4091-250MG APOSBICL4091-50MG
2-(Dimethoxymethyl)-5-(methoxymethyl)furan ≥99%
2-(Dimethoxymethyl)-5-(methoxymethyl)furan
Lagertemperatur: Raumtemperatur
CAS-Nummer: 110339-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR