(R,S)-Flurbiprofen-acyl-β-D-glucuronide ≥98%
Lieferant: Apollo Scientific
BICL4098-1MG
BICL4098-5MG
BICL4098-2MG
APOSBICL4098-1MGEA
336
EUR
APOSBICL4098-1MG
APOSBICL4098-5MG
APOSBICL4098-2MG
(R,S)-Flurbiprofen-acyl-β-D-glucuronide ≥98%
(R,S)-Flurbiprofen-acyl-β-D-glucuronide
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
91683-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...