Fenofibric acid-acyl-β-D-glucuronide ≥98%
Lieferant: Apollo Scientific
BICL4102-1MG
BICL4102-2MG
APOSBICL4102-1MGEA
434
EUR
APOSBICL4102-1MG
APOSBICL4102-2MG
Fenofibric acid-acyl-β-D-glucuronide ≥98%
Fenofibric acid-acyl-β-D-glucuronide
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
60318-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...