β-D-Glucopyranosyl-glycolaldehyde ≥95%

Lieferant: Apollo Scientific

Synonyme: 2-Oxoethyl β-D-glucopyranoside, GG

BICL4107-10MG BICL4107-25MG BICL4107-5MG
APOSBICL4107-10MGEA 532 EUR
APOSBICL4107-10MG APOSBICL4107-25MG APOSBICL4107-5MG
β-D-Glucopyranosyl-glycolaldehyde ≥95%
β-D-Glucopyranosyl-glycolaldehyde
Lagertemperatur: Raumtemperatur
CAS-Nummer: 136670-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR