L-Gulofuranuronic acid, γ-lactone ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
L-Gulofurano-6,3-lactone, L-Guluronolactone
BICL4110-10MG
BICL4110-25MG
BICL4110-5MG
APOSBICL4110-10MGEA
504
EUR
APOSBICL4110-10MG
APOSBICL4110-25MG
APOSBICL4110-5MG
L-Gulofuranuronic acid, γ-lactone ≥98%
L-Gulofuranuronic acid, γ-lactone
Schmelzpunkt:
139…141 °C Lagertemperatur: Gefrierschrank |
CAS-Nummer:
14474-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...