2,3,6,2',3',4',6'-Hepta-O-acetyl-1-O-trichloroacetimidoyl-D-cellobioside ≥97%

Lieferant: Apollo Scientific

Synonyme: 2,3,6,2',3',4',6'-Hepta-O-acetyl-D-cellobiosyl trichloroacetimidate

BICL4111-1G BICL4111-500MG
APOSBICL4111-1GEA 978 EUR
APOSBICL4111-1G APOSBICL4111-500MG
2,3,6,2',3',4',6'-Hepta-O-acetyl-1-O-trichloroacetimidoyl-D-cellobioside ≥97%
2,3,6,2',3',4',6'-Hepta-O-acetyl-1-O-trichloroacetimidoyl-D-cellobioside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 250330-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR