2,3-O-Isopropylidene-D-erythrofuranose ≥98%

Lieferant: Apollo Scientific

Synonyme: D-Erythrose acetonide

BICL4118-1G BICL4118-500MG BICL4118-2G
APOSBICL4118-1GEA 754 EUR
APOSBICL4118-1G APOSBICL4118-500MG APOSBICL4118-2G
2,3-O-Isopropylidene-D-erythrofuranose ≥98%
2,3-O-Isopropylidene-D-erythrofuranose
Lagertemperatur: Raumtemperatur
CAS-Nummer: 189996-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR