(R,S)-Ketoprofen-acyl-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4124-1MG BICL4124-2MG
APOSBICL4124-1MGEA 475 EUR
APOSBICL4124-1MG APOSBICL4124-2MG
(R,S)-Ketoprofen-acyl-β-D-glucuronide ≥95%
(R,S)-Ketoprofen-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 76690-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR