Mefenamic acid-acyl-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific
Achtung

BICL4129-1MG BICL4129-2MG
APOSBICL4129-1MGEA 586 EUR
APOSBICL4129-1MG APOSBICL4129-2MG
Mefenamic acid-acyl-β-D-glucuronide ≥95%
Mefenamic acid-acyl-β-D-glucuronide
Formel: C₂₁H₂₃NO₈
Molecular Weight: 417,409 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09840854
CAS-Nummer: 102623-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR