4-Methoxyphenyl 2,3,6-tri-O-acetyl-β-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific

BICL4134-250MG BICL4134-500MG
APOSBICL4134-250MGEA 768 EUR
APOSBICL4134-250MG APOSBICL4134-500MG
4-Methoxyphenyl 2,3,6-tri-O-acetyl-β-D-glucopyranoside ≥98%
4-Methoxyphenyl 2,3,6-tri-O-acetyl-β-D-glucopyranoside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1004771-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR