4-Methoxyphenyl 2,3,6-tri-O-benzoyl-β-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxyphenyl β-D-glucopyranoside 2,3,6-tribenzoate

BICL4138-1G BICL4138-500MG BICL4138-2G
APOSBICL4138-1GEA 560 EUR
APOSBICL4138-1G APOSBICL4138-500MG APOSBICL4138-2G
4-Methoxyphenyl 2,3,6-tri-O-benzoyl-β-D-glucopyranoside ≥98%
4-Methoxyphenyl 2,3,6-tri-O-benzoyl-β-D-glucopyranoside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 578007-30-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR