4-Methylumbelliferyl β-D-cellohexaoside ≥95%
Lieferant: Apollo Scientific
Synonyme:
4-MU β-D-cellohexaoside
BICL4153-1MG
BICL4153-2MG
APOSBICL4153-1MGEA
949
EUR
APOSBICL4153-1MG
APOSBICL4153-2MG
4-Methylumbelliferyl β-D-cellohexaoside ≥95%
4-Methylumbelliferyl β-D-cellohexaoside
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
84325-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...