Methyl 4,6-O-benzylidene-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4,6-O-benzylidene-N-Cbz-α-D-glucosaminide
BICL4156-1G
BICL4156-500MG
BICL4156-250MG
APOSBICL4156-1GEA
1270
EUR
APOSBICL4156-1G
APOSBICL4156-500MG
APOSBICL4156-250MG
Methyl 4,6-O-benzylidene-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%
Methyl 4,6-O-benzylidene-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
60076-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...