Methyl 3-O-benzyl-4,6-O-benzylidene-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-O-benzyl-4,6-O-benzylidene-N-Cbz-α-D-glucosaminide

BICL4157-1G BICL4157-250MG BICL4157-500MG
APOSBICL4157-1GEA 1550 EUR
APOSBICL4157-1G APOSBICL4157-250MG APOSBICL4157-500MG
Methyl 3-O-benzyl-4,6-O-benzylidene-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%
Methyl 3-O-benzyl-4,6-O-benzylidene-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 87907-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR