Methyl 6-O-acetyl-3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%

Lieferant: Apollo Scientific

Synonyme: Methyl 6-O-acetyl-3-O-benzyl-N-Cbz-α-D-glucosaminide

BICL4159-100MG BICL4159-25MG BICL4159-50MG
APOSBICL4159-100MGEA 1550 EUR
APOSBICL4159-100MG APOSBICL4159-25MG APOSBICL4159-50MG
Methyl 6-O-acetyl-3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%
Methyl 6-O-acetyl-3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 114869-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR