Methyl 3-O-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-β-D-galactopyranoside ≥98%
Lieferant: Apollo Scientific
Synonyme:
β-D-GlcNAc(1-3)-β-D-Gal-1-OMe
BICL4168-5MG
BICL4168-1MG
BICL4168-2MG
APOSBICL4168-5MGEA
782
EUR
APOSBICL4168-5MG
APOSBICL4168-1MG
APOSBICL4168-2MG
Methyl 3-O-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-β-D-galactopyranoside ≥98%
Methyl 3-O-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-β-D-galactopyranoside
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
93253-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...