D-Mannofuranuronic acid, γ-lactone ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: D-Mannuronolactone, D-Mannurono-6,3-lactone

BICL4173-100MG BICL4173-50MG BICL4173-250MG
APOSBICL4173-100MGEA 560 EUR
APOSBICL4173-100MG APOSBICL4173-50MG APOSBICL4173-250MG
D-Mannofuranuronic acid, γ-lactone ≥98%
D-Mannofuranuronic acid, γ-lactone
Schmelzpunkt: 141…143 °C
Lagertemperatur: Gefrierschrank
CAS-Nummer: 575-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR