Niacin-acyl-β-D-glucuronide ≥95%
Lieferant: Apollo Scientific
Synonyme:
Vitamin B3-acyl-β-D-glucuronide, Nicotinic acid-acyl-β-D-glucuronide
BICL4180-1MG
BICL4180-5MG
BICL4180-2MG
APOSBICL4180-1MGEA
351
EUR
APOSBICL4180-1MG
APOSBICL4180-5MG
APOSBICL4180-2MG
Niacin-acyl-β-D-glucuronide ≥95%
Niacin-acyl-β-D-glucuronide
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
24719-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...