N-(n-Nonyl)-1-deoxynojirimycin ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
NN-DNJ
BICL4182-10MG
BICL4182-25MG
BICL4182-5MG
APOSBICL4182-10MGEA
462
EUR
APOSBICL4182-10MG
APOSBICL4182-25MG
APOSBICL4182-5MG
N-(n-Nonyl)-1-deoxynojirimycin ≥99%
N-(n-Nonyl)-1-deoxynojirimycin
Formel:
C₁₅H₃₁NO₄ Molecular Weight: 289,411 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00905846 CAS-Nummer: 81117-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...