2-Nitrophenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-cellobioside ≥98%

Lieferant: Apollo Scientific

Synonyme: o-Nitrophenyl β-D-cellobioside heptaacetate

BICL4184-250MG BICL4184-500MG BICL4184-100MG
APOSBICL4184-250MGEA 434 EUR
APOSBICL4184-250MG APOSBICL4184-500MG APOSBICL4184-100MG
2-Nitrophenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-cellobioside ≥98%
2-Nitrophenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-cellobioside
Lagertemperatur: Raumtemperatur
CAS-Nummer: 70867-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR