Valproic acid-acyl-D-glucuronide ≥98%
Lieferant: Apollo Scientific
Achtung
Synonyme: Valproate-acyl-D-glucuronide
BICL4186-1MG BICL4186-5MG BICL4186-2MG
APOSBICL4186-1MGEA 405 EUR
APOSBICL4186-1MG APOSBICL4186-5MG APOSBICL4186-2MG
Valproic acid-acyl-D-glucuronide ≥98%
Valproic acid-acyl-D-glucuronide
Formel: C₁₄H₂₄O₈
Molecular Weight: 320.336 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00869967
CAS-Nummer: 60113-83-9